(4-Acetamidophenyl)boronsäure

Lieferant: COMBI-BLOCKS

BB-2124-1G BB-2124-5G
COBBBB-2124-1GEA 50.8 EUR
COBBBB-2124-1G COBBBB-2124-5G
(4-Acetamidophenyl)boronsäure
(4-Acetamidophenyl)boronsäure
Formel: C₈H₁₀BNO₃
MDL: MFCD02179451
CAS-Nummer: 101251-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR