Alkaliblau (C.I. 42765)-Lösung Indikator, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.09198.1000 1.09198.2500
1.09198.1000EA 128 EUR
1.09198.1000 1.09198.2500
Alkaliblau (C.I. 42765)-Lösung Indikator, Supelco®
Alkaliblau (C.I. 42765)
Formel: C₃₇H₂₉N₃O₃S
Molecular Weight: 595,73 g/mol
Siedepunkt: 80 °C (1013 hPa)
Dichte: 0,84 g/cm3 (20 °C)
Flammpunkt: 4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083221
CAS-Nummer: 1324-76-1
UN: 1993
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

dentity (UV/VIS-Spectrum) passes test
Density (d 20 °C/ 4 °C) 0.836 - 0.838
Absorption maximum λmax. (1 : 10 diluted) 603 - 607 nm
Absorbance A (λmax; 1 : 10 diluted; 1 cm) ≥ 1.00
Suitability as indicator (according to DIN 51558) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR