Fmoc-L-phenylalanin

Lieferant: NOVABIOCHEM

Synonyme: N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine

8.52011.0250 8.52016.0250
8.52011.0250EA 158 EUR
8.52011.0250 8.52016.0250
Fmoc-L-phenylalanin
Fmoc-L-phenylalanin
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387.4 g/mol
MDL: MFCD00037128
CAS-Nummer: 35661-40-6
EINECS: 252-661-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR