4-Brom-2-(piperidin-1-ylsulfonyl)anisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-((5-Bromo-2-methoxyphenyl)sulphonyl)piperidine

OT-0199-5G OT-0199-1G
COBBOT-0199-5GEA 0 EUR
COBBOT-0199-5G COBBOT-0199-1G
4-Brom-2-(piperidin-1-ylsulfonyl)anisol
4-Brom-2-(piperidin-1-ylsulfonyl)anisol
Formel: C₁₂H₁₆BrNO₃S
MDL: MFCD02043682
CAS-Nummer: 295360-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR