4-Brom-2-(pyrrolidin-1-ylsulfonyl)anisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-((5-Bromo-2-methoxyphenyl)sulphonyl)pyrrolidine

OT-0305-5G OT-0305-1G
COBBOT-0305-5GEA 303 EUR
COBBOT-0305-5G COBBOT-0305-1G
4-Brom-2-(pyrrolidin-1-ylsulfonyl)anisol
4-Brom-2-(pyrrolidin-1-ylsulfonyl)anisol
Formel: C₁₁H₁₄BrNO₃S
Molecular Weight: 320.2000 g/mol
MDL: MFCD04091911
CAS-Nummer: 691381-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR