1-Phenyl-1H-pyrazol-4-carbaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde
H63695.MD H63695.03
H63695.MDEA 56.3 EUR
H63695.MD H63695.03
1-Phenyl-1H-pyrazol-4-carbaldehyd ≥98%
1-Phenyl-1H-pyrazol-4-carbaldehyd
Purity: 98% CAS No.: 54605-72-0 Molecular Formula: C10H8N2O Formula Weight: 172.18 Physical Form: Solid Sensitivity: Moisture and Light Sensitive Appearance: Pale yellow Melting Point: 84-88° MDL No.: MFCD02179568
Formel: C₁₀H₈N₂O
Molecular Weight: 172.18 g/mol
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179568
CAS-Nummer: 54605-72-0