4'-Brom-2'-(trifluormethoxy)acetanilid

Lieferant: COMBI-BLOCKS

AN-1575-5G AN-1575-25G
COBBAN-1575-5GEA 49.7 EUR
COBBAN-1575-5G COBBAN-1575-25G
4'-Brom-2'-(trifluormethoxy)acetanilid
4'-Brom-2'-(trifluormethoxy)acetanilid
Formel: C₉H₇BrF₃NO₂
MDL: MFCD00203979
CAS-Nummer: 175278-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR