Kalium(4-butoxyphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3225-1G TB-3225-25G TB-3225-5G
COBBTB-3225-1GEA 127 EUR
COBBTB-3225-1G COBBTB-3225-25G COBBTB-3225-5G
Kalium(4-butoxyphenyl)trifluoroborat
Potassium-4-butoxyphenyltrifluoroborate
Formel: C₁₀H₁₃BF₃KO
Molecular Weight: 256.12 g/mol
MDL: MFCD04115755
CAS-Nummer: 850623-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR