3-Methoxy-N,N-dimethylanilin

Lieferant: CHEMPUR

SEW05702.250 SEW05702.1
CHMPSEW05702.250EA 0 EUR
CHMPSEW05702.250 MAYBSEW05702.1
3-Methoxy-N,N-dimethylanilin
3-Methoxy-N,N-dimethylanilin
Formel: C₉H₁₃NO
MDL: MFCD00051779
CAS-Nummer: 15799-79-8
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR