Kalium(3-fluor-4-phenylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3220-1G TB-3220-25G TB-3220-5G
COBBTB-3220-1GEA 0 EUR
COBBTB-3220-1G COBBTB-3220-25G COBBTB-3220-5G
Kalium(3-fluor-4-phenylphenyl)trifluoroborat
Potassium-3-fluoro-4-phenylphenyltrifluoroborate
Formel: C₁₂H₈BF₄K
MDL: MFCD08235035
CAS-Nummer: 850623-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR