1,3-Dibrom-5-tert-butylbenzol
Lieferant: COMBI-BLOCKS
Synonyme:
1,3-Dibromo-5-(tert-butyl)benzene, 5-(tert-Butyl)-1,3-dibromobenzene
OT-0629-1G
OT-0629-25G
COBBOT-0629-1GEA
22.7
EUR
COBBOT-0629-1G
COBBOT-0629-25G
1,3-Dibrom-5-tert-butylbenzol
1,3-Dibrom-5-tert-butylbenzol
Formel:
C₁₀H₁₂Br₂ Molecular Weight: 292,0100 g/mol |
MDL:
MFCD06657956 CAS-Nummer: 129316-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...