Benzylchloromethylether ≥70%, tech.
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
((Chloromethoxy)methyl)benzene
H60207.AD
H60207.AK
H60207.ADEA
106
EUR
H60207.AD
H60207.AK
Benzylchloromethylether ≥70%, tech.
Benzylchloromethylether
Formel:
C₈H₉ClO Molecular Weight: 156,61 g/mol Siedepunkt: 102 °C Dichte: 1,126 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000886 CAS-Nummer: 3587-60-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...