2-Chlor-5-methylanisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Chloro-2-methoxy-4-methylbenzene

OT-0708-5G OT-0708-25G OT-0708-1G
COBBOT-0708-5GEA 312 EUR
COBBOT-0708-5G COBBOT-0708-25G COBBOT-0708-1G
2-Chlor-5-methylanisol
2-Chlor-5-methylanisol
Formel: C₈H₉ClO
MDL: MFCD01679710
CAS-Nummer: 73909-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR