3-Chlor-N,N-dimethylanilin
Lieferant: CHEMPUR
SEW05670.250
SEW05670.1
CHMPSEW05670.250EA
0
EUR
CHMPSEW05670.250
MAYBSEW05670.1
3-Chlor-N,N-dimethylanilin
3-Chlor-N,N-dimethylanilin
Formel:
C₈H₁₀ClN Molecular Weight: 155,63 g/mol |
MDL:
MFCD00018456 CAS-Nummer: 6848-13-1 EINECS: 229-935-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...