1H,1H,2H,2H-Perfluoroctyltrimethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane, Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane, Trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane

SILS13160.5 SILS13160.25
SILNSILS13160.5EA 0 EUR
SILNSILS13160.5 SILNSILS13160.25
1H,1H,2H,2H-Perfluoroctyltrimethoxysilan
1H,1H,2H,2H-Perfluoroctyltrimethoxysilan
Formel: C₁₁H₁₃F₁₃O₃Si
Molecular Weight: 468,2800 g/mol
MDL: MFCD00153261
CAS-Nummer: 85857-16-5
EINECS: 288-657-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR