6-(Trifluormethyl)nicotinamid

Lieferant: FLUOROCHEM

009365-1G 009365-5G
FLUO009365-1GEA 31.1 EUR
FLUO009365-1G FLUO009365-5G
6-(Trifluormethyl)nicotinamid
6-(Trifluormethyl)nicotinamid
Formel: C₇H₅F₃N₂O
Molecular Weight: 190,1200 g/mol
MDL: MFCD01862646
CAS-Nummer: 386715-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR