1-(2-Tetrahydropyranyl)-1H-pyrazol
Lieferant: COMBI-BLOCKS
Synonyme:
1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole
HI-1494-5G
HI-1494-1G
COBBHI-1494-5GEA
38
EUR
COBBHI-1494-5G
COBBHI-1494-1G
1-(2-Tetrahydropyranyl)-1H-pyrazol
1-(2-Tetrahydropyranyl)-1H-pyrazol
Formel:
C₈H₁₂N₂O Molecular Weight: 152,20 g/mol |
MDL:
MFCD11044692 CAS-Nummer: 449758-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...