2-Brom-4'-(dimethylamino)acetophenon
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Dimethylamino)phenacyl bromide, 2-Bromo-1-(4-(dimethylamino)phenyl)ethanone
OT-0388-5G
OT-0388-1G
COBBOT-0388-5GEA
0
EUR
COBBOT-0388-5G
COBBOT-0388-1G
2-Brom-4'-(dimethylamino)acetophenon
2-Brom-4'-(dimethylamino)acetophenon
Formel:
C₁₀H₁₂BrNO |
MDL:
MFCD06409221 CAS-Nummer: 37904-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...