4-(4-Chlorphenyl)-1-buten-4-ol ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-(4-Chlorophenyl)-3-buten-1-ol, 1-(4-Chlorophenyl)but-3-en-1-ol
H53381.03
H53381.06
H53381.03EA
39.6
EUR
H53381.03
H53381.06
4-(4-Chlorphenyl)-1-buten-4-ol ≥97%
4-(4-Chlorphenyl)-1-buten-4-ol
Purity: 97%
CAS No.: 14506-33-3
Molecular Formula: C10H11ClO
Formula Weight: 182.65
Refractive Index: 1.5440
Boiling Point: 140°/2mm
MDL No.: MFCD03427246
Formel:
C₁₀H₁₁ClO Molecular Weight: 182.65 g/mol Siedepunkt: 140 °C (2 mmHg) Dichte: 1.137 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427246 CAS-Nummer: 14506-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...