4-(2,2,2-Trifluorethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((2,2,2-Trifluoroethyl)carbamoyl)phenyl)boronic acid

BB-3024-1G BB-3024-5G
COBBBB-3024-1GEA 243 EUR
COBBBB-3024-1G COBBBB-3024-5G
4-(2,2,2-Trifluorethylcarbamoyl)phenylboronsäure
4-(2,2,2-Trifluorethylcarbamoyl)phenylboronsäure
Formel: C₉H₉BF₃NO₃
Molecular Weight: 246,9800 g/mol
MDL: MFCD08235087
CAS-Nummer: 874459-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR