1H-Benzimidazol-5-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

296783-25G 296783-5G
SIAL296783-25GEA 188 EUR
SIAL296783-25G SIAL296783-5G
1H-Benzimidazol-5-carbonsäure, Sigma-Aldrich®
1H-Benzimidazol-5-carbonsäure
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD00011555
CAS-Nummer: 15788-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR