3-(Furfurylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-(Furfurylcarbamoyl)benzeneboronic acid
BB-3069-5G
BB-3069-1G
COBBBB-3069-5GEA
678
EUR
COBBBB-3069-5G
COBBBB-3069-1G
3-(Furfurylcarbamoyl)phenylboronsäure
3-(Furfurylcarbamoyl)phenylboronsäure
Formel:
C₁₂H₁₂BNO₄ Molecular Weight: 245,04 g/mol |
MDL:
MFCD04115700 CAS-Nummer: 850567-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...