6-Cyan-7-nitrochinoxalin-2,3-dion ≥98%

Lieferant: Cayman Chemical

Synonyme: 7-Nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6-Cyano-7-nitroquinoxaline-2,3(1H,4H)-dione, CNQX

14618-5 14618-10 14618-25 14618-50
CAYM14618-5EA 67 EUR
CAYM14618-5 CAYM14618-10 CAYM14618-25 CAYM14618-50
6-Cyan-7-nitrochinoxalin-2,3-dion ≥98%
6-Cyan-7-nitrochinoxalin-2,3-dion

CNQX is a competitive, non-NMDA glutamate receptor antagonist (IC50s = 0.3 and 1.5 μM for AMPA and kainate receptors, respectively, versus IC50 = 25 μM for NMDA receptors).
This compound has been used to specifically target AMPA and kainate receptor responses and thus differentiate from that of NMDA receptors.

Formel: C₉H₄N₄O₄
Molecular Weight: 232,16 g/mol
MDL: MFCD00069232
CAS-Nummer: 115066-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR