Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazol-4-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Ethyl 5-amino-1-(p-tolyl)-1H-pyrazole-4-carboxylate

4W-0243.500 4W-0243.1 4W-0243.5 4W-0243.10
BIOT4W-0243.500EA 0 EUR
BIOT4W-0243.500 BIOT4W-0243.1 BIOT4W-0243.5 BIOT4W-0243.10
Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazol-4-carboxylat
Formel: C₁₃H₁₅N₃O₂
Molecular Weight: 245.2800 g/mol
MDL: MFCD00973936
CAS-Nummer: 15001-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR