3-(Cyclopentylcarbamoyl)phenylboronsäureneopentylglycolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Cyclopentyl-3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzamide

NE-3064-5G NE-3064-1G
COBBNE-3064-5GEA 0 EUR
COBBNE-3064-5G COBBNE-3064-1G
3-(Cyclopentylcarbamoyl)phenylboronsäureneopentylglycolester
3-(Cyclopentylcarbamoyl)phenylboronsäureneopentylglycolester
Formel: C₁₇H₂₄BNO₃
MDL: MFCD06659914
CAS-Nummer: 850567-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR