2-(Diethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Diethylcarbamoyl)phenyl)boronic acid

BB-2951-5G BB-2951-1G
COBBBB-2951-5GEA 1930 EUR
COBBBB-2951-5G COBBBB-2951-1G
2-(Diethylcarbamoyl)phenylboronsäure
2-(Diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221.0600 g/mol
MDL: MFCD01318988
CAS-Nummer: 129112-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR