2-(Dimethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Dimethylcarbamoyl)benzeneboronic acid

BB-2950-5G BB-2950-1G
COBBBB-2950-5GEA 1520 EUR
COBBBB-2950-5G COBBBB-2950-1G
2-(Dimethylcarbamoyl)phenylboronsäure
2-(Dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193,01 g/mol
MDL: MFCD03425959
CAS-Nummer: 874219-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR