3-(Diethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N,N-Diethylaminocarbonyl)phenylboronic acid, 3-(Diethylcarbamoyl)benzeneboronic acid, (3-(Diethylcarbamoyl)phenyl)boronic acid

BB-3051-5G BB-3051-25G BB-3051-1G
COBBBB-3051-5GEA 0 EUR
COBBBB-3051-5G COBBBB-3051-25G COBBBB-3051-1G
3-(Diethylcarbamoyl)phenylboronsäure
3-(Diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
MDL: MFCD04115684
CAS-Nummer: 237413-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR