3-(Dimethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N,N-Dimethylaminocarbonyl)phenylboronic acid, 3-(Dimethylcarbamoyl)benzeneboronic acid, (3-(Dimethylcarbamoyl)phenyl)boronic acid

BB-3050-5G BB-3050-25G BB-3050-1G
COBBBB-3050-5GEA 183 EUR
COBBBB-3050-5G COBBBB-3050-25G COBBBB-3050-1G
3-(Dimethylcarbamoyl)phenylboronsäure
3-(Dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193,01 g/mol
MDL: MFCD03412066
CAS-Nummer: 373384-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR