4-(Diethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid

BB-3001-5G BB-3001-25G BB-3001-1G
COBBBB-3001-5GEA 75.9 EUR
COBBBB-3001-5G COBBBB-3001-25G COBBBB-3001-1G
4-(Diethylcarbamoyl)phenylboronsäure
4-(Diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221,06 g/mol
MDL: MFCD03411949
CAS-Nummer: 389621-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR