3-(Isobutylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Isobutylaminocarbonyl)phenylboronic acid

BB-3061-5G BB-3061-25G BB-3061-1G
COBBBB-3061-5GEA 142 EUR
COBBBB-3061-5G COBBBB-3061-25G COBBBB-3061-1G
3-(Isobutylcarbamoyl)phenylboronsäure
3-(Isobutylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
MDL: MFCD04115692
CAS-Nummer: 723282-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR