3-(tert-Butylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(tert-Butylaminocarbonyl)phenylboronic acid

BB-3062-25G BB-3062-1G BB-3062-5G
COBBBB-3062-25GEA 1180 EUR
COBBBB-3062-25G COBBBB-3062-1G COBBBB-3062-5G
3-(tert-Butylcarbamoyl)phenylboronsäure
3-(tert-Butylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
MDL: MFCD04115693
CAS-Nummer: 183158-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR