3-(Methylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid
BB-3056-5G
BB-3056-1G
COBBBB-3056-5GEA
0
EUR
COBBBB-3056-5G
COBBBB-3056-1G
3-(Methylcarbamoyl)phenylboronsäure
3-(Methylcarbamoyl)phenylboronsäure
Formel:
C₈H₁₀BNO₃ |
MDL:
MFCD04038918 CAS-Nummer: 832695-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...