3-(Propylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N-Propylaminocarbonyl)benzeneboronic acid, 3-(N-Propylaminocarbonyl)phenylboronic acid

BB-3058-5G BB-3058-1G
COBBBB-3058-5GEA 451 EUR
COBBBB-3058-5G COBBBB-3058-1G
3-(Propylcarbamoyl)phenylboronsäure
3-(Propylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
MDL: MFCD04038919
CAS-Nummer: 850567-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR