4-(Butylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Butylaminocarbonyl)phenylboronic acid

BB-3010-5G BB-3010-25G BB-3010-1G
COBBBB-3010-5GEA 152 EUR
COBBBB-3010-5G COBBBB-3010-25G COBBBB-3010-1G
4-(Butylcarbamoyl)phenylboronsäure
4-(Butylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
MDL: MFCD03788412
CAS-Nummer: 252663-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR