4-(Ethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(N-Ethylaminocarbonyl)phenylboronic acid

BB-3007-1G BB-3007-5G
COBBBB-3007-1GEA 147 EUR
COBBBB-3007-1G COBBBB-3007-5G
4-(Ethylcarbamoyl)phenylboronsäure
4-(Ethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193.0100 g/mol
MDL: MFCD03411954
CAS-Nummer: 850568-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR