4-(Methylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Methylcarbamoyl)benzeneboronic acid, 4-(N-Methylaminocarbonyl)phenylboronic acid

BB-3006-1G BB-3006-5G
COBBBB-3006-1GEA 0 EUR
COBBBB-3006-1G COBBBB-3006-5G
4-(Methylcarbamoyl)phenylboronsäure
4-(Methylcarbamoyl)phenylboronsäure
Formel: C₈H₁₀BNO₃
MDL: MFCD03788427
CAS-Nummer: 121177-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR