4-(Propylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Propylcarbamoyl)phenyl)boronic acid, 4-(n-Propylcarbamoyl)benzeneboronic acid

BB-3008-5G BB-3008-1G
COBBBB-3008-5GEA 451 EUR
COBBBB-3008-5G COBBBB-3008-1G
4-(Propylcarbamoyl)phenylboronsäure
4-(Propylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
MDL: MFCD06656015
CAS-Nummer: 171922-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR