3-(Benzylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N-Benzylaminocarbonyl)phenylboronic acid

BB-3055-1G BB-3055-5G BB-3055-25G
COBBBB-3055-1GEA 98.3 EUR
COBBBB-3055-1G COBBBB-3055-5G COBBBB-3055-25G
3-(Benzylcarbamoyl)phenylboronsäure
3-(Benzylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₄BNO₃
MDL: MFCD04115688
CAS-Nummer: 625470-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR