Methyl-2-chlor-6-methylisonicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-chloro-6-picoline-4-carboxylate, Methyl 2-chloro-6-methylpyridine-4-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

649481-1G 649481-5G
SIAL649481-1GEA 38.4 EUR
SIAL649481-1G SIAL649481-5G
Methyl-2-chlor-6-methylisonicotinat, Sigma-Aldrich®
Methyl-2-chlor-6-methylisonicotinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
MDL: MFCD04038391
CAS-Nummer: 3998-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR