3-(Chlormethyl)-5-phenyl-1,2,4-oxadiazol

Lieferant: FLUOROCHEM

017400-2.5G 017400-10G 017400-25G 017400-1G
FLUO017400-2.5GEA 0 EUR
FLUO017400-2.5G FLUO017400-10G FLUO017400-25G FLUO017400-1G
3-(Chlormethyl)-5-phenyl-1,2,4-oxadiazol
3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole
Formel: C₉H₇ClN₂O
Molecular Weight: 194,6177 g/mol
MDL: MFCD00084968
CAS-Nummer: 1201-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR