3-Methyl-5-isoxazolcarbonsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Methylisoxazole-5-carboxylic acid

H63740.MD H63740.03
H63740.MDEA 81.4 EUR
H63740.MD H63740.03
3-Methyl-5-isoxazolcarbonsäure ≥97%
3-Methyl-5-isoxazolecarboxylic acid

Purity: 97% CAS No.: 4857-42-5 Molecular Formula: C5H5NO3 Formula Weight: 127.10 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00464222

Formel: C₅H₅NO₃
Molecular Weight: 127,10 g/mol
MDL: MFCD00464222
CAS-Nummer: 4857-42-5
EINECS: 225-454-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR