2-Amino-3-brombenzonitril 95%
Lieferant: Thermo Scientific
|
Achtung
|
H61823.MD
H61823.03
H61823.MDEA
98.3
EUR
H61823.MD
H61823.03
2-Amino-3-brombenzonitril 95%
2-Amino-3-brombenzonitril
Formel:
C₇H₅BrN₂ Molecular Weight: 197,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05670539 CAS-Nummer: 114344-60-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...