3-(Trifluormethyl)isonicotinsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)pyridine-4-carboxylic acid

H63174.03 H63174.06
H63174.03EA 117 EUR
H63174.03 H63174.06
3-(Trifluormethyl)isonicotinsäure 95%
3-(Trifluormethyl)isonicotinsäure

Purity: 95% CAS No.: 590371-38-3 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Physical Form: Solid Appearance: Off-white MDL No.: MFCD08234940

Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234940
CAS-Nummer: 590371-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR