Methyl-2-amino-3-methylbenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 2-amino-3-methylbenzoate

11X-0097.1 11X-0097.10 11X-0097.5 11X-0097.500
BIOT11X-0097.1EA 0 EUR
BIOT11X-0097.1 BIOT11X-0097.10 BIOT11X-0097.5 BIOT11X-0097.500
Methyl-2-amino-3-methylbenzoat
Methyl-2-amino-3-methylbenzoat
Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
MDL: MFCD06200918
CAS-Nummer: 22223-49-0
EINECS: 244-847-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR