Ethyl-4-hydroxy-6-(trifluormethoxy)chinolin-3-carboxylat

Lieferant: FLUOROCHEM

008310-1G 008310-5G
FLUO008310-1GEA 30 EUR
FLUO008310-1G FLUO008310-5G
Ethyl-4-hydroxy-6-(trifluormethoxy)chinolin-3-carboxylat
Ethyl-4-hydroxy-6-(trifluormethoxy)chinolin-3-carboxylat
Formel: C₁₃H₁₀F₃NO₄
MDL: MFCD00219847
CAS-Nummer: 175203-85-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR