Ethyl-6-fluor-4-hydroxychinolin-3-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate

5J-031.500 5J-031.1 5J-031.5 5J-031.10
BIOT5J-031.500EA 0 EUR
BIOT5J-031.500 BIOT5J-031.1 BIOT5J-031.5 BIOT5J-031.10
Ethyl-6-fluor-4-hydroxychinolin-3-carboxylat
Ethyl-6-fluor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀FNO₃
Molecular Weight: 235.22 g/mol
MDL: MFCD00173345
CAS-Nummer: 318-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR