2-(Trifluormethyl)-4-chinolinol

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Trifluoromethyl)quinolin-4-ol, 4-Hydroxy-2-(trifluoromethyl)quinoline

HC-2164-5G HC-2164-1G
COBBHC-2164-5GEA 74.2 EUR
COBBHC-2164-5G COBBHC-2164-1G
2-(Trifluormethyl)-4-chinolinol
2-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,1560 g/mol
MDL: MFCD00153196
CAS-Nummer: 1701-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR