2-(4-Bromphenyl)-4,7-dichlorchinazolin

Lieferant: CHEMPUR

79-0045.500 79-0045.1
CHMP79-0045.500EA 0 EUR
CHMP79-0045.500 CHMP79-0045.1
2-(4-Bromphenyl)-4,7-dichlorchinazolin
2-(4-Bromophenyl)-4,7-dichloroquinazoline
Formel: C₁₄H₇BrCl₂N₂
MDL: MFCD04115263
CAS-Nummer: 405933-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR