2'-Brom-4'-nitroacetanilid

Lieferant: COMBI-BLOCKS

AN-1444-1G AN-1444-5G
COBBAN-1444-1GEA 0 EUR
COBBAN-1444-1G COBBAN-1444-5G
2'-Brom-4'-nitroacetanilid
2'-Brom-4'-nitroacetanilid
Formel: C₈H₇BrN₂O₃
MDL: MFCD00569284
CAS-Nummer: 57045-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR